TrackIt loading buffers are 6X solutions used for easy loading and tracking of DNA samples in agarose gels. They containRead more
Have Questions?
Catalog Number
Quantity
10482035
3 x 0.5 mL
Catalog number 10482035
Price (USD)
71.65
Online exclusive
74.00
Save 2.35
Each
Quantity:
3 x 0.5 mL
Request bulk or custom format
Price (USD)
71.65
Online exclusive
74.00
Save 2.35
Each
TrackIt loading buffers are 6X solutions used for easy loading and tracking of DNA samples in agarose gels. They contain two tracking dyes: one that runs behind the sample and one that runs ahead of the sample. The tracking dyes serve as visual markers of migration during electrophoresis and indicate when maximum resolution of the DNA fragments has been achieved. Additionally, visualization of DNA bands is not obscured by the tracking dyes because they run outside the limits of most DNA samples.TrackIt Cyan/Orange Loading Buffer is recommended for DNA fragments between 50 bp and 10 Kb.TrackIt Cyan/Yellow Loading Buffer is recommended for DNA fragments between 10 bp and 1 Kb.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration6X
Gel CompatibilityAgarose Gels
Quantity3 x 0.5 mL
Sample Loading Volume0.5 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Gel TypeAgarose
Label or DyeTartrazine, Xylene Cyanol, Xylene Cyanol FF
Product LineTrackIt
Product TypeLoading Buffer
Unit SizeEach
Contents & Storage
• 3 x 0.5 mL 6X TrackIt Cyan/Yellow Loading Buffer
Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.