p-Anisidine, 99%
p-Anisidine, 99%
p-Anisidine, 99%
Thermo Scientific Chemicals

p-Anisidine, 99%

CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 104830010
Price (EUR)
91,60
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
91,60
Each
Chemical Identifiers
CAS104-94-9
IUPAC Name4-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyBHAAPTBBJKJZER-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or granules or flakes
Melting point56°C to 59°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only