Anisoin, 95%
Anisoin, 95%
Anisoin, 95%
Thermo Scientific Chemicals

Anisoin, 95%

CAS: 119-52-8 | C16H16O4 | 272.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 104840250
Price (USD)
56.50
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
56.50
Each
Chemical Identifiers
CAS119-52-8
IUPAC Name2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one
Molecular FormulaC16H16O4
InChI KeyLRRQSCPPOIUNGX-UHFFFAOYNA-N
SMILESCOC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Color)White to yellow
Appearance (Form)Powder
Melting point108°C to 115°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only