p-Anisoyl chloride, 99%
p-Anisoyl chloride, 99%
p-Anisoyl chloride, 99%
Thermo Scientific Chemicals

p-Anisoyl chloride, 99%

CAS: 100-07-2 | C8H7ClO2 | 170.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number 104850050
Price (USD)
28.65
Online exclusive
32.00
Save 3.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
28.65
Online exclusive
32.00
Save 3.35
Each
Chemical Identifiers
CAS100-07-2
IUPAC Name4-methoxybenzoyl chloride
Molecular FormulaC8H7ClO2
InChI KeyMXMOTZIXVICDSD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
GC>=98.5 %
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
Refractive index1.5800 to 1.5820 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only