TrackIt™ 100 bp DNA Ladder
Invitrogen™

TrackIt™ 100 bp DNA Ladder

Invitrogen TrackIt 100 bp DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.
Have Questions?
카탈로그 번호수량
1048805850 μg
카탈로그 번호 10488058
제품 가격(KRW)
204,000
온라인 행사
Ends: 30-Sep-2026
226,000
할인액 22,000 (10%)
Each
수량:
50 μg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
204,000
온라인 행사
Ends: 30-Sep-2026
226,000
할인액 22,000 (10%)
Each

Invitrogen TrackIt 100 bp DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.

TrackIt 100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.

Highlights of TrackIt 100 bp DNA Ladder:

  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months
  • Ready-to-use—premixed with 6X TrackIt Cyan/Orange Loading Buffer for tracking of DNA migration
  • Precise—an exact amount of DNA in each band

Product use

The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
사양
농도0.1 μg/μL
젤 호환성Agarose gel
반응 수100 Applications
수량50 μg
로드 준비Yes
샘플 로딩 부피1 mL
유통 기한24 Months when stored at room temperature.
배송 조건Approved for shipment at Room Temperature or on Dry Ice
기술Individual chromatography-purified DNA fragments
부피(미터법)50 μg
젤 유형Agarose
제품라인TrackIt
제품 유형DNA Ladder
크기 범위100 to 2000 bp
Unit SizeEach
구성 및 보관
• 500 μL TrackIt 100 bp DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 24 months. For longer periods, store at -20°C.

자주 묻는 질문(FAQ)

Can I know the sequences of Invitrogen DNA ladders?

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?

Invitrogen DNA ladders contain linear dsDNA fragments.

Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

I'm seeing anomalous migration of my DNA ladder. What happened?

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

What is the difference between the TrackIt DNA Ladders and other DNA ladders?

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.

인용 및 참조 문헌 (10)

인용 및 참조 문헌
Abstract
Characterization of genes encoding for acquired bacitracin resistance in Clostridium perfringens.
Authors:Charlebois A, Jalbert LA, Harel J, Masson L, Archambault M
Journal:PLoS One
PubMed ID:22970221
'Phenotypic bacitracin resistance has been reported in Clostridium perfringens. However, the genes responsible for the resistance have not yet been characterized. Ninety-nine C. perfringens isolates recovered from broilers and turkeys were tested for phenotypic bacitracin resistance. Bacitracin MIC(90) (>256 µg/ml) was identical for both turkey and chicken isolates; whereas MIC(50) ... More
Detection of murine leukemia virus in the Epstein-Barr virus-positive human B-cell line JY, using a computational RNA-Seq-based exogenous agent detection pipeline, PARSES.
Authors:Lin Z, Puetter A, Coco J, Xu G, Strong MJ, Wang X, Fewell C, Baddoo M, Taylor C, Flemington EK
Journal:J Virol
PubMed ID:22238296
'Many cell lines commonly used for biological studies have been found to harbor exogenous agents such as the human tumor viruses Epstein-Barr virus (EBV) and human papillomavirus. Nevertheless, broad-based, unbiased approaches to globally assess the presence of ectopic organisms within cell model systems have not previously been available. We reasoned ... More
Antibiotic-resistant Enterococcus faecalis in abattoir pigs and plasmid colocalization and cotransfer of tet(M) and erm(B) genes.
Authors:Tremblay CL, Letellier A, Quessy S, Daignault D, Archambault M
Journal:J Food Prot
PubMed ID:22947466
'This study was conducted to determine plasmid colocalization and transferability of both erm(B) and tet(M) genes in Enterococcus faecalis isolates from abattoir pigs in Canada. A total of 124 E. faecalis isolates from cecal contents of abattoir pigs were examined for antibiotic susceptibility. High percentages of resistance to macrolides and ... More
Rescue of recent virulent and avirulent field strains of bluetongue virus by reverse genetics.
Authors:van Gennip RG, van de Water SG, Potgieter CA, Wright IM, Veldman D, van Rijn PA
Journal:PLoS One
PubMed ID:22363444
'Since 1998, Bluetongue virus (BTV)-serotypes 1, 2, 4, 9, and 16 have invaded European countries around the Mediterranean Basin. In 2006, a huge BT-outbreak started after incursion of BTV-serotype 8 (BTV8) in North-Western Europe. More recently, BTV6 and BTV11 were reported in North-Western Europe in 2008. These latter strains are ... More
TGF-a/HA complex promotes tympanic membrane keratinocyte migration and proliferation via ErbB1 receptor.
Authors:Mei Teh B, Redmond SL, Shen Y, Atlas MD, Marano RJ, Dilley RJ
Journal:Exp Cell Res
PubMed ID:23384599
'Tympanic membrane perforations are common and represent a management challenge to clinicians. Current treatments for chronic perforations involve a graft surgery and require general anaesthesia, including associated costs and morbidities. Bioactive molecules (e.g. growth factors, cytokines) play an important role in promoting TM wound healing following perforation and the use ... More