This product will be discontinued on September 15, 2017. Learn more and find replacements at thermofisher.com/ladderupgrade17

E-Gel™ 25 bp DNA Ladder This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  10488095

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ready-to-use E-Gel® 25 bp DNA Ladder is premixed with loading buffer and is formulated specifically for maximum performance on E-Gel® EX gels as well as other E-Gel® precast gels. It is recommended for use with E-Gel® EX 4% gels.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Gel Compatibility: E-Gel™
Ready to Load: Yes
Concentration: 50 ng⁄µl
Product Line: E-Gel®
Product Size: 100 applications
Size Range: 25 to 500 bp
System: E-Gel®
Green Features: Sustainable packaging
Shipping Condition: Approved for shipment at Room Temperature or on Dry Ice

Contents & storage

E-Gel® 25 bp DNA Ladder is supplied at 50 ng⁄µl in buffer containing 0.0094% XCFF and 0.06% tartrazine. Store E-Gel® 25 bp DNA Ladder at 4°C. This ladder is guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products