E-Gel™ Ultra Low Range DNA Ladder
Invitrogen™

E-Gel™ Ultra Low Range DNA Ladder

Invitrogen E-Gel Ultra Low Range DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the rangeLeia mais
Have Questions?
Número do catálogoQuantity
10488096100 applications
Número do catálogo 10488096
Preço (BRL)
2.025,62
Each
Quantity:
100 applications
Preço (BRL)
2.025,62
Each
Invitrogen E-Gel Ultra Low Range DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 10 bp to 300 bp. E-Gel Ultra Low Range DNA Ladder consists of nine individual chromatography-purified DNA fragments with a reference band at 100 bp for easy orientation.

E-Gel Ultra Low Range DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel Ultra Low Range DNA Ladder:
• Performance—optimized for use on 4% E-Gel agarose gels
• Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature up to six months
• Ready to use—premixed with loading buffer
• Convenient—provided with 1X E-Gel Sample Loading Buffer
• Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 4% E-Gel agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Especificações
Concentration0.025 μg/μL
Gel CompatibilityDouble Comb E-Gels, Single Comb E-Gels
Quantity100 applications
Ready to LoadYes
Sample Loading Volume1.25 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Volume (Metric)2 x 1000 μL
Gel TypeAgarose
Product TypeDNA Ladder
Size Range10 to 300 bp
Unit SizeEach
Conteúdo e armazenamento
• 2 x 1 mL E-Gel Ultra Low Range DNA Ladder
• 1.25 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods store at -20°C.

Frequently asked questions (FAQs)

Can I know the sequences of Invitrogen DNA ladders?

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?

Invitrogen DNA ladders contain linear dsDNA fragments.

Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

I'm seeing anomalous migration of my DNA ladder. What happened?

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

What is the difference between the TrackIt DNA Ladders and other DNA ladders?

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.