Invitrogen E-Gel 50 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range ofRead more
Have Questions?
Catalog Number
Quantity
10488099
100 applications
Catalog number 10488099
Price (USD)
279.65
Online exclusive
291.00
Save 11.35
Each
Quantity:
100 applications
Price (USD)
279.65
Online exclusive
291.00
Save 11.35
Each
Invitrogen E-Gel 50 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 bp to 2,500 bp. E-Gel 50 bp DNA Ladder consists of 17 individual chromatography-purified DNA fragments and has reference bands at 2500, 800, and 350 bp for easy orientation.
E-Gel 50 bp DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.
Highlights of E-Gel 50 bp DNA Ladder: • Performance—optimal for use on E-Gel agarose gels • Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months • Ready-to-use—premixed with loading buffer • Convenient—provided with 1X E-Gel Sample Loading Buffer • Precise—an exact amount of DNA in each band
Product use The double-stranded DNA ladder can be visualized on 2% E-Gel agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.025 μg/μL
Gel CompatibilityE-Gel™ agarose gels
Quantity100 applications
Ready to LoadYes
Sample Loading Volume1 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Volume (Metric)2 x 1000 μL
Gel TypeAgarose
Product TypeDNA Ladder
Size Range50 to 2500 bp
System TypeE-Gel
Unit SizeEach
Contents & Storage
• 2 x 1 mL E-Gel 50 bp DNA Ladder • 1 mL 1X E-Gel Sample Loading Buffer
Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.
Frequently asked questions (FAQs)
Can I know the sequences of Invitrogen DNA ladders?
Sequences of Invitrogen DNA and RNA ladders are proprietary.
Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?
Invitrogen DNA ladders contain linear dsDNA fragments.
Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?
Invitrogen DNA ladders are composed of double-stranded DNA fragments only.
I'm seeing anomalous migration of my DNA ladder. What happened?
This can happen if the marker was heated. Please ensure that the ladders are not heated before use.
What is the difference between the TrackIt DNA Ladders and other DNA ladders?
TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.
TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.
The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.
Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.