Anthranilamide, 99+%
Anthranilamide, 99+%
Anthranilamide, 99+%
Thermo Scientific Chemicals

Anthranilamide, 99+%

CAS: 88-68-6 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 104901000
Price (USD)
42.60
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
42.60
Each
Chemical Identifiers
CAS88-68-6
IUPAC Name2-aminobenzamide
Molecular FormulaC7H8N2O
InChI KeyPXBFMLJZNCDSMP-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown to grey-brown
HPLC>=99.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
Melting point111°C to 114°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only