Ácido antranílico, +98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido antranílico, +98 %, Thermo Scientific Chemicals

CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
5 kg
Número de catálogo 104911000
Precio (EUR)
-
Cantidad:
100 g
Pedido a granel o personalizado
Identificadores químicos
CAS118-92-3
IUPAC Name2-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyRWZYAGGXGHYGMB-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
HPLC>=98.0 %
Infrared spectrumConforms
Appearance (Color)Light yellow to beige
Melting point144°C to 148°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only