2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
Thermo Scientific Chemicals

2-Aminobenzonitrile, 98%

CAS: 1885-29-6 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 104920100
Price (EUR)
24,70
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,70
Each
Chemical Identifiers
CAS1885-29-6
IUPAC Name2-aminobenzonitrile
Molecular FormulaC7H6N2
InChI KeyHLCPWBZNUKCSBN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige to brown
Infrared spectrumConforms
Appearance (Form)Powder or crystals or flakes and/or chunks
Melting point46°C to 51°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only