Anthrone, ACS reagent
Anthrone, ACS reagent
Anthrone, ACS reagent
Thermo Scientific Chemicals

Anthrone, ACS reagent

CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 104960250
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Chemical Identifiers
CAS90-44-8
IUPAC Name9,10-dihydroanthracen-9-one
Molecular FormulaC14H10O
InChI KeyRJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point152°C to 157°C
Sensitivity(to carbohydrates) Passes test
Absorbance=<0.75 (425 nm, 1 cm cell)
Appearance (Color)Almost white to yellow to yellow-green
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only