L(+)-Aspartic acid, 98+%
L(+)-Aspartic acid, 98+%
L(+)-Aspartic acid, 98+%
Thermo Scientific Chemicals

L(+)-Aspartic acid, 98+%

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Drum
Catalog number 105040100
Price (USD)
-
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS56-84-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<200 ppm
Sulfate (SO4)=<300 ppm
Ammonium (NH4)=<200 ppm
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only