Interested in a bulk or custom version of this product? Request pricing.

CSF2RB, Recombinant Human Protein, His Tag

Catalog number: 10516H08H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Colony Stimulating Factor 2 Receptor, Beta, Low-Affinity (CSF2RB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 443) of human CSF2RB (NP_000386.1) fused to a polyhistidine-tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA.
1. Immobilized human CD131 at 10 µg/ml (100 µl/well) can bind biotinylated human EPOR/Fc with a linear range of 0.16-4 µg/ml.
2. Immobilized human CD131 at 10 µg/ml (100 µl/well) can bind human GMCSF with a linear range of 0.032-4 µg/ml.

Formulation:140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins, Cytokines & Receptors
Research Category: Cardiovascular Research, Immunology, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Gene Aliases: CSF2RB, CD131, CDw131, IL3RB, IL5RB
Gene ID: 1439
Gene Symbol: CSF2RB
Molecular Weight: 50 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Recombinant, Truncated
Protein Name: Colony stimulating factor 2 receptor, beta, low-affinity
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of protein in suspension buffer. Store at -5 to -30°C.

Documents

Manuals & protocols

Recommended products