Benzamidine Hydrochloride Hydrate, 98%
Benzamidine Hydrochloride Hydrate, 98%
Benzamidine Hydrochloride Hydrate, 98%
Thermo Scientific Chemicals

Benzamidine Hydrochloride Hydrate, 98%

CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 105245000
Price (USD)
743.65
Online Exclusive
826.00
Save 82.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
743.65
Online Exclusive
826.00
Save 82.35 (10%)
Each
Chemical Identifiers
CAS206752-36-5
IUPAC Namebenzenecarboximidamide hydrochloride
Molecular FormulaC7H9ClN2
InChI KeyLZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILESCl.NC(=N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 % (on dry substance)
Infrared spectrumConforms
Water=<19 % (K.F.)
Appearance (Form)Powder
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only