Interested in a bulk or custom version of this product? Request pricing.

RAC1 Recombinant Human Protein, N-GST Tag

Catalog number: 10535H09B5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ras-Related C3 Botulinum Toxin Substrate 1 Protein (RAC 1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human Rac1 isoform A (NP_008839.2) (Met 1-Cys 189) fused to the GST tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: 50 mM Tris, 100 mM NaCl, 1mM GSH, 10% Glycerol, pH 8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Signal Transduction, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: RAC1, MGC111543, MIG5, TC-25, p21-Rac1
Gene ID: 5879
Gene Symbol: RAC 1
Molecular Weight: 47 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Full Length, Recombinant
Protein Name: Ras-Related C3 Botulinum Toxin Substrate 1 Protein
Protein Tag or Fusion: GST Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols