Interested in a bulk or custom version of this product? Request pricing.

FGFR4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag

Catalog number: 10538H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fibroblast growth factor receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies..

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 precursor (NP_002002.3) fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts. The ED50 for this effect is typically 2-6 ng/ml.

Formulation: Lyophilized in PBS, pH7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Cardiovascular Research, Immunology, Neurobiology, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: FGFR4, CD334, JTK2, MGC20292, TKF
Gene ID: 2264
Gene Symbol: FGFR4
Kinase Family: FGF Receptor Kinase Family
Molecular Weight: 66 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Recombinant, Truncated
Protein Name: Fibroblast growth factor receptor 4
Protein Subtype: FGFs (Fibroblast Growth Factors), Receptor Tyrosine Kinases
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products