Interested in a bulk or custom version of this product? Request pricing.

FGFR4, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag

Catalog number: 10538H03H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Fibroblast Growth Factor Receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) precursor fused to the C-terminal polyhistidine tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to inhibit FGF acidic dependent proliferation of Balb3T3 mouse fibroblasts. The ED50 for this effect is typically 2-6 ng/ml.

Formulation:140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.


Purity: ≥ 85% by SDS-PAGE
Gene ID: 2264
Species: Human
Expression: Mammalian
Gene Symbol: FGFR4
Gene Aliases: FGFR4, CD334, JTK2, MGC20292, TKF
Product Size: 5 x 50 µg
Protein Form: Truncated, Recombinant
Purification: PAGE
Protein Family: Cytokines & Receptors
Endotoxin Level: < 1 EU/µg
Molecular Weight: 66 kDa
Research Category: Immunology, Stem Cell Research, Neurobiology, Signal Transduction, Cardiovascular Research, Oncology
Protein Tag or Fusion: hIgG1-Fc His
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of protein in suspension buffer. Store at -5 to -30°C.


Manuals & protocols

Recommended products