Benzoic anhydride, 98%
Benzoic anhydride, 98%
Benzoic anhydride, 98%
Thermo Scientific Chemicals

Benzoic anhydride, 98%

CAS: 93-97-0 | C14H10O3 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105500250
Price (EUR)
19,65
Online exclusive
21,50
Save 1,85 (9%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
19,65
Online exclusive
21,50
Save 1,85 (9%)
Each
Chemical Identifiers
CAS93-97-0
IUPAC Namebenzoyl benzoate
Molecular FormulaC14H10O3
InChI KeyCHIHQLCVLOXUJW-UHFFFAOYSA-N
SMILESO=C(OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=97.5 %
Melting point38°C to 45°C
Appearance (Color)White to cream to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only