Benzoin, 98%
Benzoin, 98%
Benzoin, 98%
Thermo Scientific Chemicals

Benzoin, 98%

CAS: 119-53-9 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105510025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS119-53-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Off-white to light yellow
Melting point133°C to 137°C
Sulfated ash=<0.2 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only