Interested in a bulk or custom version of this product? Request pricing.

BMPR2 Recombinant Human Protein (without catalytic activity domain), His tag

Catalog number: 10551H08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Bone morphogenetic protein receptor type II (BMPR2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies..

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 151) of human BMPR2 (NP_001195.2) fused to a polyhistidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in PBS, pH7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Cardiovascular Research, Immunology, Neurobiology, Oncology, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: BMPR2, BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK
Gene ID: 659
Gene Symbol: BMPR2
Molecular Weight: 15.6 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Recombinant, Truncated
Protein Name: bone morphogenetic protein receptor, type II (serine/threonine kinase)
Protein Subtype: Growth Factor Receptors, Serine⁄Threonine Kinases
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products