Interested in a bulk or custom version of this product? Request pricing.

IL2RG Recombinant Human Protein, His Tag

Catalog number: 10555H08B50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 2 Receptor Gamma (IL2RG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human IL2RG (P31785) extracellular domain (Met 1-Asn 254) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA.

Formulation: 20 mM Tris, 500 mM NaCl, pH 7.4, 10% gly.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1
Gene Symbol: IL2RG
Molecular Weight: 28.8 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Name: interleukin 2 receptor gamma
Protein Subtype: Cytokine Receptors, Interleukins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols

Recommended products