Interested in a bulk or custom version of this product? Request pricing.

DDC Recombinant Human Protein, His Tag

Catalog number: 10560H08B5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dopa Decarboxylase (DDC), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to convert the substrate 3, 4-dihydroxy L-phenylalanine (L-Dopa) to 3, 4-dihydroxyphenylethylamine (dopamine). The dopamine product is measured by its absorbance at 340 nm after derivatization with trinitrobenzene sulfonic acid. The specific activity is >1500 pmoles/min/µg.

Formulation: 50 mM Tris, 100 mM NaCl, 0.2 µg/µl BSA, pH8, 10% glycerol.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: AADC, DDC
Gene Symbol: DDC
Molecular Weight: 55 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: Dopa Decarboxylase
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Research Category: Neurobiology
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols