DMEM, low glucose, GlutaMAX™ Supplement, pyruvate
Gibco™

DMEM, low glucose, GlutaMAX™ Supplement, pyruvate

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
105670141 x 500 mL
1056702210 x 500 mL
Catalog number 10567014
Price (MXN)
-
Quantity:
1 x 500 mL
Customize this product
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

  • With: Low Glucose, Sodium Pyruvate, GlutaMAX™, Phenol Red
  • Without: HEPES

The complete formulation is available.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. Gibco™ DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, 293, Cos-7, and PC-12
Cell TypeMammalian Cells
Concentration1X
Culture EnvironmentCO2
For Use With (Application)Mammalian Cell Culture
FormatBottle
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco, GlutaMAX
Product TypeDMEM (Dulbecco's Modified Eagle Medium)
Quantity1 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutaMAX, Low Glucose, Sodium Pyruvate, Phenol Red
Without AdditivesNo HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (4)

Citations & References
Abstract
Association of Bcr-Abl with the Proto-oncogene Vav Is Implicated in Activation of the Rac-1 Pathway.
Authors: Bassermann Florian; Jahn Thomas; Miething Cornelius; Seipel Petra; Bai Ren-Yuan; Coutinho Sunita; Tybulewicz Victor L; Peschel Christian; Duyster Justus;
Journal:J Biol Chem
PubMed ID:11790798
'Vav is a guanine nucleotide exchange factor for the Rho/Rac family predominantly expressed in hematopoietic cells and implicated in cell proliferation and cytoskeletal organization. The oncogenic tyrosine kinase Bcr-Abl has been shown to activate Rac-1, which is important for Bcr-Abl induced leukemogenesis. Previous studies by Matsuguchi et al. (Matsuguchi, T., ... More
The protein SET binds the neuronal Cdk5 activator p35nck5a and modulates Cdk5/p35nck5a activity.
Authors: Qu Dianbo; Li Qing; Lim Hui-Ying; Cheung Nam Sang; Li Rong; Wang Jerry H; Qi Robert Z;
Journal:J Biol Chem
PubMed ID:11741927
'The neuronal Cdk5 kinase is composed of the catalytic subunit Cdk5 and the activator protein p35(nck5a) or its isoform, p39(nck5ai). To identify novel p35(nck5a)- and p39(nck5ai)-binding proteins, fragments of p35(nck5a) and p39(nck5ai) were utilized in affinity isolation of binding proteins from rat brain homogenates, and the isolated proteins were identified ... More
Disrupted-in-Schizophrenia-1 (DISC-1): mutant truncation prevents binding to NudE-like (NUDEL) and inhibits neurite outgrowth.
Authors:Ozeki Y, Tomoda T, Kleiderlein J, Kamiya A, Bord L, Fujii K, Okawa M, Yamada N, Hatten ME, Snyder SH, Ross CA, Sawa A,
Journal:Proc Natl Acad Sci U S A
PubMed ID:12506198
Disrupted-in-Schizophrenia-1 (DISC-1) is a gene whose mutant truncation is associated with major psychiatric illness with a predominance of schizophrenic symptomatology. We have cloned and characterized rodent DISC-1. DISC-1 expression displays pronounced developmental regulation with the highest levels in late embryonic life when the cerebral cortex develops. In yeast two-hybrid analyses, ... More
KRIT1, a gene mutated in cerebral cavernous malformation, encodes a microtubule-associated protein.
Authors: Gunel Murat; Laurans Maxwell S H; Shin Dana; DiLuna Michael L; Voorhees Jennifer; Choate Keith; Nelson-Williams Carol; Lifton Richard P;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12140362
Mutations in Krev1 interaction trapped gene 1 (KRIT1) cause cerebral cavernous malformation, an autosomal dominant disease featuring malformation of cerebral capillaries resulting in cerebral hemorrhage, strokes, and seizures. The biological functions of KRIT1 are unknown. We have investigated KRIT1 expression in endothelial cells by using specific anti-KRIT1 antibodies. By both ... More