4-Benzoylbenzoic acid, 99%
4-Benzoylbenzoic acid, 99%
4-Benzoylbenzoic acid, 99%
Thermo Scientific Chemicals

4-Benzoylbenzoic acid, 99%

CAS: 611-95-0 | C14H10O3 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 105720250
Price (EUR)
223,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
223,00
Each
Chemical Identifiers
CAS611-95-0
IUPAC Name4-benzoylbenzoic acid
Molecular FormulaC14H10O3
InChI KeyIFQUPKAISSPFTE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point197°C to 200°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only