How do you prepare blood cells for chromosome analysis?
Phytohemagglutinin Assay:
Lymphocytes are differentiated cells which normally do not undergo subsequent cell divisions. By culturing lymphocytes in the presence of a mitogen (KaryoMAX Phytohemagglutinin (M-Form) (PHA), Cat. No. 10576), they are stimulated to replicate their DNA and enter into mitosis. After an optimum time of the cells being cultured (46 h for a newborn and 68 h for an adult), a mitotic inhibitor, KaryoMAX COLCEMID Solution (Cat. No. 15210 or 15212), is added to the lymphocyte culture for 20 min. The addition of COLCEMID to dividing cells acts to prevent the synthesis of spindle fibers and, therefore, to stop mitosis in metaphase. Metaphase is the optimum phase of mitosis for microscopically visualizing the chromosomes. By submitting cells to a hypotonic solution and a series of fixation steps, metaphase chromosomes can be microscopically observed and analyzed.
As a quality control measure, each lot of Phytohemagglutinin is tested as a chromosome reagent for the examination of metaphase spreads used for cytogenetic studies. This reagent is evaluated by supplementation to an approved, non-phytohemagglutinin containing chromosome medium. These samples are then supplemented with freshly collected human peripheral blood and have been found to be acceptable in their ability to produce blastogenesis with human lymphocytes when compared to a previously tested control.
Test Procedure:
1. The required volume of peripheral blood is collected aseptically in a sodium heparinized vacutainer tube or syringe.
2. Add 10 ml of either PB-MAX Karyotyping Medium (Cat. No. 10386) to each sterile T-25 flask to be set up for the assay.
3. Add 0.75ml blood to each tube.
4. Incubate flask in CO2 incubator for 48 to 68 h with caps loose.
5. Add 0.05-0.1 ug/ml COLCEMID to each flask for a 15 minute incubation.
6. After 15 min, transfer flask contents to a 15 ml centrifuge tube and spin down at 1,200 rpm for 5 min.
7. Remove supernatant and resuspend pellet.
8. Add 10 ml of 0.068 M KCl to pellet and gently mix. Allow to sit at room temperature for 15 min. Add 0.5 ml of fixative (three parts absolute methanol to one part glacial acetic acid). Gently mix with pipette.
9. Centrifuge for 5 min at 1,200 rpm. Aspirate off supernatant. Add 10 ml of fixative, mix, and let sit at room temperature for 10 min.
10. Repeat centrifugation step. Add 5 ml of fixative, mix, and let sit for 10 min at room temperature.
11. Centrifuge and aspirate off supernatant. Add 5 ml of fixative and incubate for 10 min at 4°C.
12. Centrifuge at 1,200 rpm for 7 min. Aspirate off supernatant. Gently resuspend in fixative.
13. Prepare slide by placing 6 to 8 drops of cell suspension on slide. Use warming plate to dry slides.
14. Once all traces of moisture have disappeared, you can begin staining procedure:
- Stain with Giemsa Stain for 6 to 8 min, or any other appropriate stain.
- Remove slides from stain and rinse under running distilled water.
- Allow slides to thoroughly dry and examine slides for well-spread metaphases.
Record number of cells and at the same time record the number of these cells which are in metaphase. At least 500 cells must be counted for a valid assay. Statistically determine the Mean Mitotic Index (a quantitative measure) and Mean Banding Resolution (a qualitative measure). The test sample values must favorably compare to the reference control values.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Lymphocytes are differentiated cells which normally do not undergo subsequent cell divisions. By culturing lymphocytes in the presence of a mitogen (KaryoMAX Phytohemagglutinin (M-Form) (PHA), Cat. No. 10576), they are stimulated to replicate their DNA and enter into mitosis. After an optimum time of the cells being cultured (46 h for a newborn and 68 h for an adult), a mitotic inhibitor, KaryoMAX COLCEMID Solution (Cat. No. 15210 or 15212), is added to the lymphocyte culture for 20 min. The addition of COLCEMID to dividing cells acts to prevent the synthesis of spindle fibers and, therefore, to stop mitosis in metaphase. Metaphase is the optimum phase of mitosis for microscopically visualizing the chromosomes. By submitting cells to a hypotonic solution and a series of fixation steps, metaphase chromosomes can be microscopically observed and analyzed.
As a quality control measure, each lot of Phytohemagglutinin is tested as a chromosome reagent for the examination of metaphase spreads used for cytogenetic studies. This reagent is evaluated by supplementation to an approved, non-phytohemagglutinin containing chromosome medium. These samples are then supplemented with freshly collected human peripheral blood and have been found to be acceptable in their ability to produce blastogenesis with human lymphocytes when compared to a previously tested control.
Test Procedure:
1. The required volume of peripheral blood is collected aseptically in a sodium heparinized vacutainer tube or syringe.
2. Add 10 ml of either PB-MAX Karyotyping Medium (Cat. No. 10386) to each sterile T-25 flask to be set up for the assay.
3. Add 0.75ml blood to each tube.
4. Incubate flask in CO2 incubator for 48 to 68 h with caps loose.
5. Add 0.05-0.1 ug/ml COLCEMID to each flask for a 15 minute incubation.
6. After 15 min, transfer flask contents to a 15 ml centrifuge tube and spin down at 1,200 rpm for 5 min.
7. Remove supernatant and resuspend pellet.
8. Add 10 ml of 0.068 M KCl to pellet and gently mix. Allow to sit at room temperature for 15 min. Add 0.5 ml of fixative (three parts absolute methanol to one part glacial acetic acid). Gently mix with pipette.
9. Centrifuge for 5 min at 1,200 rpm. Aspirate off supernatant. Add 10 ml of fixative, mix, and let sit at room temperature for 10 min.
10. Repeat centrifugation step. Add 5 ml of fixative, mix, and let sit for 10 min at room temperature.
11. Centrifuge and aspirate off supernatant. Add 5 ml of fixative and incubate for 10 min at 4°C.
12. Centrifuge at 1,200 rpm for 7 min. Aspirate off supernatant. Gently resuspend in fixative.
13. Prepare slide by placing 6 to 8 drops of cell suspension on slide. Use warming plate to dry slides.
14. Once all traces of moisture have disappeared, you can begin staining procedure:
- Stain with Giemsa Stain for 6 to 8 min, or any other appropriate stain.
- Remove slides from stain and rinse under running distilled water.
- Allow slides to thoroughly dry and examine slides for well-spread metaphases.
Record number of cells and at the same time record the number of these cells which are in metaphase. At least 500 cells must be counted for a valid assay. Statistically determine the Mean Mitotic Index (a quantitative measure) and Mean Banding Resolution (a qualitative measure). The test sample values must favorably compare to the reference control values.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.