Benzoylhydrazine, 98%
Benzoylhydrazine, 98%
Benzoylhydrazine, 98%
Thermo Scientific Chemicals

Benzoylhydrazine, 98%

CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 105771000
Price (USD)
78.65
Online exclusive
86.80
Save 8.15
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
78.65
Online exclusive
86.80
Save 8.15
Each
Chemical Identifiers
CAS613-94-5
IUPAC Namebenzohydrazide
Molecular FormulaC7H8N2O
InChI KeyWARCRYXKINZHGQ-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
Melting point113°C to 117°C
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only