Interested in a bulk or custom version of this product? Request pricing.

ACVR1B (ALK4) Recombinant Human Protein (without catalytic activity domain), hIgG1-Fc His tag

Catalog number: 10583H03H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Activin A Receptor, Type Ib (ACVR1B), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

This recombinant protein was expressed from a DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) fused to C-terminal His-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human TDGF1 at 2 µg/ml (100 µl/well) can bind human ACVR1B with a linear range of 0.0068-0.16 µg/ml.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Signal Transduction
Product Line: Sino Biological®
Product Size: 50 µg

Contents & storage

Includes 1 vial of protein in suspension buffer. Store at -20°C.

Documents

Manuals & protocols

Recommended products