N-Benzylglycine ethyl ester, 96%
N-Benzylglycine ethyl ester, 96%
N-Benzylglycine ethyl ester, 96%
Thermo Scientific Chemicals

N-Benzylglycine ethyl ester, 96%

CAS: 6436-90-4 | C11H15NO2 | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 105950100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6436-90-4
IUPAC Nameethyl 2-(benzylamino)acetate
Molecular FormulaC11H15NO2
InChI KeyULOLIZHBYWAICY-UHFFFAOYSA-N
SMILESCCOC(=O)CNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5045 to 1.5065 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only