N-Benzylidenemethylamine, 99%
N-Benzylidenemethylamine, 99%
N-Benzylidenemethylamine, 99%
Thermo Scientific Chemicals

N-Benzylidenemethylamine, 99%

CAS: 622-29-7 | C8H9N | 119.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 106010250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS622-29-7
IUPAC Namemethyl(phenylmethylidene)amine
Molecular FormulaC8H9N
InChI KeyHXTGGPKOEKKUQO-UHFFFAOYSA-N
SMILESCN=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only