5-Benzyloxytryptamine hydrochloride, 98%
5-Benzyloxytryptamine hydrochloride, 98%
5-Benzyloxytryptamine hydrochloride, 98%
Thermo Scientific Chemicals

5-Benzyloxytryptamine hydrochloride, 98%

CAS: 52055-23-9 | C17H18N2O·HCl | 302.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 106112500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52055-23-9
IUPAC Name2-[5-(benzyloxy)-1H-indol-3-yl]ethan-1-amine hydrochloridyl
Molecular FormulaC17H18ClN2O
InChI KeyNIDXFJIFFPKQBY-UHFFFAOYSA-N
SMILES[Cl].NCCC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to beige
Titration Argentometric>=97.5 %
HPLC>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only