N-Benzyl-4-piperidone, 99%
N-Benzyl-4-piperidone, 99%
N-Benzyl-4-piperidone, 99%
Thermo Scientific Chemicals

N-Benzyl-4-piperidone, 99%

CAS: 3612-20-2 | C12H15NO | 189.26 g/mol
数量:
100 mL
250 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 106131000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3612-20-2
IUPAC Name1-benzylpiperidin-4-one
Molecular FormulaC12H15NO
InChI KeySJZKULRDWHPHGG-UHFFFAOYSA-N
SMILESO=C1CCN(CC2=CC=CC=C2)CC1
さらに表示
Titration with HClO498 to 102 %
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to straw yellow
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only