Cloruro de benciltrifenilfosfonio, 99 %, Thermo Scientific Chemicals
Cloruro de benciltrifenilfosfonio, 99 %, Thermo Scientific Chemicals
Cloruro de benciltrifenilfosfonio, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cloruro de benciltrifenilfosfonio, 99 %, Thermo Scientific Chemicals

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 106215000
Precio (USD)
-
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1100-88-5
IUPAC Namebenzyltriphenylphosphanium chloride
Molecular FormulaC25H22ClP
InChI KeyUSFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Para formación de enlaces C-C (Olefinación)
RUO – Research Use Only