4-Biphenylcarboxylic acid, 98%
4-Biphenylcarboxylic acid, 98%
4-Biphenylcarboxylic acid, 98%
Thermo Scientific Chemicals

4-Biphenylcarboxylic acid, 98%

CAS: 92-92-2 | C13H10O2 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 106280250
Price (USD)
49.65
Online exclusive
55.00
Save 5.35
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.00
Save 5.35
Each
Chemical Identifiers
CAS92-92-2
IUPAC Name[1,1'-biphenyl]-4-carboxylic acid
Molecular FormulaC13H10O2
InChI KeyNNJMFJSKMRYHSR-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point223°C to 228°C
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Titration with NaOH97.5 to 102.5 %
Appearance (Color)White to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only