Automation Reservoirs, 96 Channel, with removable splash baffles, Non-Sterile - FAQs
View additional product information for Automation Reservoirs - FAQs (1064-05-6, 1064-15-8, 1064-05-7, 1064-15-7, 1064-15-6, 1064-05-8)
3 product FAQs found
What is the maximum volume held by each Automation Reservoir used with the Versette Automatic Liquid Handler?
The maximum volumes for the Automation Reservoirs are as follows:
- 96-channel reservoirs hold a minimum of 95 mL w/ baffle and 125 mL w/o baffle
- 96-channel DW reservoirs hold a minimum of 160 mL w/baffle and 220 mL w/o baffle
- 384-channel reservoirs hold a minimum of 65 mL w/baffle and 95 mL w/o baffle
What are the minimum (dead) volumes for the Automation Reservoirs used with the Versette Automatic Liquid Handler?
The minimum dead volumes for the Automation Reservoirs are as follows:
- 96-channel reservoirs: 5 mL
- 384-channel reservoirs: 4 mL
Can reservoirs be automatically filled with the Versette Automated Liquid Handler?
Yes, a non-contact (N/C) reservoir fill module is included as an accessory with the purchase of a peristaltic pump (Cat. No. 650-04-PUMP).