Search
View additional product information for Automation Reservoirs - FAQs (1064-05-6, 1064-15-8, 1064-05-7, 1064-15-7, 1064-15-6, 1064-05-8)
3 product FAQs found
The maximum volumes for the Automation Reservoirs are as follows:
- 96-channel reservoirs hold a minimum of 95 mL w/ baffle and 125 mL w/o baffle
- 96-channel DW reservoirs hold a minimum of 160 mL w/baffle and 220 mL w/o baffle
- 384-channel reservoirs hold a minimum of 65 mL w/baffle and 95 mL w/o baffle
The minimum dead volumes for the Automation Reservoirs are as follows:
- 96-channel reservoirs: 5 mL
- 384-channel reservoirs: 4 mL
Yes, a non-contact (N/C) reservoir fill module is included as an accessory with the purchase of a peristaltic pump (Cat. No. 650-04-PUMP).