L-Bornyl acetate, 95%
L-Bornyl acetate, 95%
L-Bornyl acetate, 95%
Thermo Scientific Chemicals

L-Bornyl acetate, 95%

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol
Quantity:
5 g
100 g
500 g
Catalog number 106551000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS5655-61-8
IUPAC Name(1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Molecular FormulaC12H20O2
InChI KeyKGEKLUUHTZCSIP-HOSYDEDBSA-N
SMILESCC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation(20°C, 589 nm) (neat) < -38°
Appearance (Form)Liquid
Specific gravity(15/15°C) 0.986 to 0.99
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only