4'-Bromoacetophenone, 98%
4'-Bromoacetophenone, 98%
4'-Bromoacetophenone, 98%
Thermo Scientific Chemicals

4'-Bromoacetophenone, 98%

CAS: 99-90-1 | C8H7BrO | 199.05 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 106590250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS99-90-1
IUPAC Name1-(4-bromophenyl)ethan-1-one
Molecular FormulaC8H7BrO
InChI KeyWYECURVXVYPVAT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(Br)C=C1
さらに表示
GC>=97.5 %
Melting point48°C to 52°C
Appearance (Form)Crystals or flakes
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only