3-Bromoanisole, 99+%
3-Bromoanisole, 99+%
3-Bromoanisole, 99+%
Thermo Scientific Chemicals

3-Bromoanisole, 99+%

CAS: 2398-37-0 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 106621000
Price (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2398-37-0
IUPAC Name1-bromo-3-methoxybenzene
Molecular FormulaC7H7BrO
InChI KeyPLDWAJLZAAHOGG-UHFFFAOYSA-N
SMILESCOC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only