Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Thermo Scientific Chemicals

Bromobenzene, 99%, pure

CAS: 108-86-1 | C6H5Br | 157.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1066800252.5 LGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 106680025
Price (USD)
245.65
Online Exclusive
273.00
Save 27.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
245.65
Online Exclusive
273.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS108-86-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5580 to 1.5600 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only