Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Thermo Scientific Chemicals

Bromobenzene, 99%, pure

CAS: 108-86-1 | C6H5Br | 157.01 g/mol
数量:
250 mL
1 L
2.5 L
10 L
パッケージ:
すべて表示
Plastic Drum
製品番号(カタログ番号) 106680100
価格(JPY)
-
数量:
10 L
パッケージ:
Plastic Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-86-1
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5580 to 1.5600 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only