Interested in a bulk or custom version of this product? Request pricing.

XCL1 Recombinant Human Protein, His Tag

Catalog number: 10672H08B5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Chemokine (C Motif) Ligand 1 (XCL1), recombinant human protein is supplied in a frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human XCL1 (P47992) (Met 1- Gly 114) fused to a polyhistidine tag at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in 20 mM, Tris 500 mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 10% glycerol.

Reconstitution: No reconstitution is needed. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: ATAC, LPTN, LTN, SCM-1, SCM-1a, SCM1, SCM1A, SCYC1
Gene ID: 6375
Gene Symbol: XCL1
Molecular Weight: 11.6 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Chemokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Chemokine (C motif) ligand 1
Protein Subtype: Other Cytokines
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: > 85% by SDS-PAGE
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols