2-Bromobenzoyl chloride, 98%
2-Bromobenzoyl chloride, 98%
2-Bromobenzoyl chloride, 98%
Thermo Scientific Chemicals

2-Bromobenzoyl chloride, 98%

CAS: 7154-66-7 | C7H4BrClO | 219.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 106750500
Price (EUR)
211,00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
211,00
Each
Chemical Identifiers
CAS7154-66-7
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5945 to 1.5985 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to orange
Appearance (Form)Liquid
Infrared spectrumConforms
Titration RCOCl97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only