Search
| Número do catálogo | Type | For Use With (Equipment) |
|---|---|---|
| 1067531 | Organic Matrix Accessory | Element Series HR-ICP-MS |
| 1159440 | Matrix Kit | Element Series HR-ICP-MS |
| 1162170 | Acid-Resistant Sample Intro | Element Series HR-ICP-MS |
| 1324711 | Organic Matrix Kit | iCAP MX and iCAP Q/Qnova |
| 1324720 | Acid Resistant Kit Q/Qnova | iCAP Q/Qnova ICP-MS |
| BRE0064559 | iCAP MX Acid Resistance Kit | iCAP MX Series ICP-MS |
Enhance your analysis with dedicated Thermo Scientific™ Sample Handling Kits for the Element Series HR-ICP-MS, the iCAP MX and iCAP Q/Qnova Series ICP-MS instruments.
Enable the routine analysis of organic solvents, aggressively acidic or highly complex sample matrices with these convenient, complete sample handling accessory kits.
Enhance System Performance with Tailored Sampling Kits
Organic Matrix Accessory Kit for Element Series HR-ICP-MS
Organic solvents are easily handled with this kit comprising:
Acid-Resistant Sample Introduction System
For the routine analysis of aggressive sample matrices, this kit includes:
Complex Matrix Sample Handling Kit for Element Series HR-ICP-MS
Enable the routine analysis of highly complex matrices with this product bundle containing: