1-Bromobutane, 99%
1-Bromobutane, 99%
1-Bromobutane, 99%
Thermo Scientific Chemicals

1-Bromobutane, 99%

CAS: 109-65-9 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 106770500
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
30.30
Save 4.65
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
30.30
Save 4.65
Each
Chemical Identifiers
CAS109-65-9
IUPAC Name1-bromobutane
Molecular FormulaC4H9Br
InChI KeyMPPPKRYCTPRNTB-UHFFFAOYSA-N
SMILESCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4380 to 1.4410 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only