4-Bromo-3,5-dimethylphenol, 99%
4-Bromo-3,5-dimethylphenol, 99%
4-Bromo-3,5-dimethylphenol, 99%
Thermo Scientific Chemicals

4-Bromo-3,5-dimethylphenol, 99%

CAS: 7463-51-6 | C8H9BrO | 201.06 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
產品號碼 106870100
價格 (TWD)
-
Quantity:
10 g
Request bulk or custom format
化學識別
CAS7463-51-6
IUPAC Name4-bromo-3,5-dimethylphenol
Molecular FormulaC8H9BrO
InChI KeyWMUWDPLTTLJNPE-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC(C)=C1Br
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals
Appearance (Color)Colorless to light pink
Melting point113°C to 116°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only