1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
Thermo Scientific Chemicals

1-Bromododecane, 98%

CAS: 143-15-7 | C12H25Br | 249.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 106912500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS143-15-7
IUPAC Name1-bromododecane
Molecular FormulaC12H25Br
InChI KeyPBLNBZIONSLZBU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<125 APHA
Infrared spectrumConforms
Water=<0.1 % (K.F.)
GC>=97.5 %
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only