CSF2RA (GMCSFR) Recombinant Human Protein (His Tag)

Catalog number: 10701H08H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Colony stimulating factor 2 receptor alpha (CSF2RA/GMCSFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 320) of human CSF2RA (NP_034100.2) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to inhibit CSF2 dependent proliferation of TF-1 human erythroleukemic cells. The EC50 for this effect is typically 3-5 µg/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

Specifications

Research Category: Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD116, Csfgmra, GM-CSFR, GM-CSF-Ra, Csf2ra
Gene Symbol: CSF2RA
Molecular Weight: 36 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Cytokines & Receptors
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage)
Protein Subtype: Colony Stimulating Factors
Protein Tag or Fusion: His Tag
Purity: > 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products