5-Bromofuroic acid, 99%
5-Bromofuroic acid, 99%
5-Bromofuroic acid, 99%
Thermo Scientific Chemicals

5-Bromofuroic acid, 99%

CAS: 585-70-6 | C5H3BrO3 | 190.98 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 107020250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS585-70-6
IUPAC Name5-bromofuran-2-carboxylic acid
Molecular FormulaC5H3BrO3
InChI KeyYVTQHZDUDUCGRD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Br)O1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to cream to beige to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
Melting point188°C to 192°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only