1-Bromohexane, 99+%
1-Bromohexane, 99+%
1-Bromohexane, 99+%
Thermo Scientific Chemicals

1-Bromohexane, 99+%

CAS: 111-25-1 | C6H13Br | 165.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
107045000500 mLGlass bottle
Catalog number 107045000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-25-1
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Appearance (Form)Clear liquid
GC>=99.0 %
Refractive index1.4465 to 1.4485 (20°C, 589 nm)
GC=<0.5 % Hexanol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only